Learn More
ATCC YUMM5.2; Melanoma; Mouse (Mus musculus)
Catalog No. 502382661
Encompass
YUMM5.2 is a cell line with epithelial-like morphology isolated in 2015 from the skin of an adult male mouse with melanoma. This cell line is genetically modified containing human relevant mutations like BrafV600E (wild-type); p53-/-. It also has Tyrosinase:CreERT2 transgene which restricts recombination to melanocytes. It was deposited by the Yale University School of Medicine.
Catalog No. 50-238-2661 Supplier ATCC Supplier No. CRL3367
YUMM5.2 is a cell line with epithelial-like morphology isolated in 2015 from the skin of an adult male mouse with melanoma. This cell line is genetically modified containing human relevant mutations like BrafV600E (wild-type); p53-/-. It also has Tyrosinase:CreERT2 transgene which restricts recombination to melanocytes. It was deposited by the Yale University School of Medicine.