Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ YUMMER1.7-H2B-GFP5 Mouse Melanoma Cell Line

Catalog No. SCC245MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 x 10&sup6; Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC245MI 1 x 10&sup6; Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC245MI Supplier MilliporeSigma™ Supplier No. SCC245
Add to Cart
Add to Cart

YUMMER1.7-H2B-GFP5 mouse melanoma cell line for studies of immune checkpoint inhibition and mechanisms of antitumor responses

  • Both immunocompetent and reflective of the somatic mutations common in melanomas
  • YUMMER1.7 H2B-GFP carries three driver mutations of melanoma:
    • Braf V600E
    • Pten -/-
    • Cdkn2 -/- (1,2)
  • Harbors a high frequency of stable UV-induced somatic mutations which have shown to stimulate host adaptive immune response

Contains genetically modified organisms (GMO)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Cells can be cultured for 10 passages after initial thawing without significantly affecting cell marker expression and functionality
Quantity 1 x 10&sup6; Cells/Vial
For Use With (Application) Cell Culture
Sample Type Cancer cells

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.