Learn More
Cayman Chemical YunaconItIn 10mg
SDP
Catalog No. 502946853
Encompass_Preferred
A diterpenoid alkaloid with diverse biological activities; active against E. coli and B. subtilis (MICs = 32 and 8 UG/ml, respectively); inhibits ferrozine-Fe2+ complex formation (IC50 = 11.6 UG/ml); ; has been found in the urine of patients with aconite poisoning
Catalog No. 50-294-6853 Supplier Cayman Chemical Supplier No. 3412710 MG
A diterpenoid alkaloid with diverse biological activities; active against E. coli and B. subtilis (MICs = 32 and 8 UG/ml, respectively); inhibits ferrozine-Fe2+ complex formation (IC50 = 11.6 UG/ml); ; has been found in the urine of patients with aconite poisoning