Learn More
CPC Scientific H-Tyr-Val-Ala-Asp-CHO 1MG
SDP
Supplier:  CPC Scientific CASP032A
Encompass
ONE-LETTER SEQUENCE: YVAD-CHOMOLECULAR FORMULA: C21H30N4O7MOLECULAR WEIGHT:450.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: A. Suzuki, Exp. Cell Res., 234, 507 (1997)
Catalog No. 50-210-8978