Learn More
Cayman Chemical YVAD-CHO (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502941038
Encompass_Preferred
A caspase-1/ICE inhibitor; prevents ICE-like and CPP32-like proteinase activity and inhibits apoptosis induced by ceramide or irinotecan in 4B1 cells at 600 µM; prevents chromogranin A-induced cell death of primary rat microglia at 100 µM
Catalog No. 50-294-1038 Supplier Cayman Chemical Supplier No. 274205 MG
A caspase-1/ICE inhibitor; prevents ICE-like and CPP32-like proteinase activity and inhibits apoptosis induced by ceramide or irinotecan in 4B1 cells at 600 µM; prevents chromogranin A-induced cell death of primary rat microglia at 100 µM