Learn More
Aviva Systems Biology Corp YWHAE Antibody - middle region (AVARP02058_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02058_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YWHAE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human YWHAE. Alternate names: 14-3-3E, FLJ45465, KCIP-1, MDCR, MDSApplicationWB. Species reactivity: Dog, Guinea Pig, Mouse, Zebrafish
Catalog No. 50-014-62496