Learn More
Aviva Systems Biology Corp YWHAH Antibody - N-terminal region (AVARP02061_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02061_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YWHAH. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human YWHAH. Alternate names: YWHA1ApplicationIHC, WB. Species reactivity: Dog, Human, Rabbit, Zebrafish
Catalog No. 50-014-62497