Learn More
Medchemexpress LLC Yx-02-030 | 3049076-98-1 | 99.9% | 1267.41 | C66H85Cl2N9O10S | 10 MG
SDP
Catalog No. 5000288690
Encompass_Preferred
A PROTAC small-molecule degrader that targets MDM2 for research use in biochemical and cell-based assays. It binds MDM2 and recruits the VHL E3 ubiquitin ligase to induce proteasomal degradation of MDM2, disrupting MDM2-p53 interactions.
  • High purity (≈99.9%).
  • Available as a 10 mg research pack suitable for assay development.
  • Potent inhibition of MDM2-p53 binding and VHL engagement in biochemical assays.
  • Molecular weight 1267.41; formula C66H85Cl2N9O10S.
  • Provided with an SDS for handling and storage guidance.
Catalog No. 50-002-88690 Supplier Medchemexpress LLC Supplier No. HY15868410MG
A PROTAC small-molecule degrader that targets MDM2 for research use in biochemical and cell-based assays. It binds MDM2 and recruits the VHL E3 ubiquitin ligase to induce proteasomal degradation of MDM2, disrupting MDM2-p53 interactions.
  • High purity (≈99.9%).
  • Available as a 10 mg research pack suitable for assay development.
  • Potent inhibition of MDM2-p53 binding and VHL engagement in biochemical assays.
  • Molecular weight 1267.41; formula C66H85Cl2N9O10S.
  • Provided with an SDS for handling and storage guidance.