Learn More
Aviva Systems Biology Corp YY1 Antibody - N-terminal region (ARP38301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human YY1. Alternate names: DELTA, NF-E1, UCRBP, INO80S, YIN-YANG-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Sheep, Zebrafish
Catalog No. 50-014-56601