Learn More
Santa Cruz Biotechnology YY1 Antibody (H-10), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC7341
Encompass
YY1 Antibody (H-10) is a high quality monoclonal YY1 antibody suitable for the detection of the YY1 protein of mouse, rat and human origin. YY1 Antibody (H-10) is available as both the non-conjugated anti-YY1 antibody form, as well as multiple conjugated forms of anti-YY1 antibody, including agarose, HRP, PE, FITC and multiple Alexa Fluor conjugates.
Catalog No. NC1422701