Learn More
Aviva Systems Biology Corp YY1AP1 Antibody - middle region (ARP50779_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50779_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YY1AP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human YY1AP1. Alternate names: FLJ10875, FLJ13914, HCCA1, HCCA2, YAP, YY1APApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-285-539