Learn More
Medchemexpress LLC Yya-021 | 144217-65-2 | 99.0% | 317.43 g/mol | C18H27N3O2 | 25 MG
SDP
Catalog No. 5000215721
Encompass_Preferred
YYA-021 is a small-molecule CD4 mimic that inhibits HIV entry by competitively blocking the viral envelope glycoprotein interaction with CD4. It exhibits potent anti-HIV activity with low cytotoxicity and is supplied for research applications as a high-purity solid.
  • Small-molecule CD4 mimic that blocks HIV entry.
  • Demonstrates high anti-HIV activity with low cytotoxicity.
  • High purity (≈99%) suitable for biochemical and cellular assays.
  • Supplied as a white to off-white solid with multiple pack sizes.
  • Storage recommendations support long-term stability for research use.
Catalog No. 50-002-15721 Supplier Medchemexpress LLC Supplier No. HY10003925MG
YYA-021 is a small-molecule CD4 mimic that inhibits HIV entry by competitively blocking the viral envelope glycoprotein interaction with CD4. It exhibits potent anti-HIV activity with low cytotoxicity and is supplied for research applications as a high-purity solid.
  • Small-molecule CD4 mimic that blocks HIV entry.
  • Demonstrates high anti-HIV activity with low cytotoxicity.
  • High purity (≈99%) suitable for biochemical and cellular assays.
  • Supplied as a white to off-white solid with multiple pack sizes.
  • Storage recommendations support long-term stability for research use.