Learn More
Medchemexpress LLC YZ9 | 6093-71-6 | 98.9% | 234.21 | 50 MG
SDP
Catalog No. 5000343915
Encompass_Preferred
YZ9 is a potent PFKFB3 inhibitor with an IC50 of 0.183 μM, competitively inhibiting Fru-6-P with a Ki value of 0.094 μM. It demonstrates antitumor effects against cervical cancer.
  • Potent PFKFB3 inhibitor
  • Exhibits antitumor effects against cervical cancer
Catalog No. 50-003-43915 Supplier Medchemexpress LLC Supplier No. HY11015650MG
YZ9 is a potent PFKFB3 inhibitor with an IC50 of 0.183 μM, competitively inhibiting Fru-6-P with a Ki value of 0.094 μM. It demonstrates antitumor effects against cervical cancer.
  • Potent PFKFB3 inhibitor
  • Exhibits antitumor effects against cervical cancer