Learn More
Medchemexpress LLC (Z)-1-((2-(trifluoromethyl)phenyl)methylene)-5-(thiazol-2-yl)thiourea | 438190-29-5 | MFCD01152003 | 99.9% | 273.23 g·mol⁻¹ | C11H6F3NO2S | 10 MG
SDP
Catalog No. 5000289234
Encompass_Preferred
SMI-4a is a selective, cell-permeable, ATP-competitive inhibitor of Pim-1 kinase supplied as a high-purity research chemical. It inhibits Pim-1 (IC50 = 24 μM; Ki = 0.6 μM) and Pim-2 (IC50 = 100 μM), is provided as a white to off-white solid in a 10 mg package, and includes recommended storage conditions for powder and solutions.
  • Selective ATP-competitive Pim-1 kinase inhibitor suitable for research use.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a 10 mg solid for small-scale experiments.
  • Includes storage recommendations for powder and dissolved samples.
  • Contains chemical identifiers for unambiguous compound tracking.
Catalog No. 50-002-89234 Supplier Medchemexpress LLC Supplier No. HY16576A10MG
SMI-4a is a selective, cell-permeable, ATP-competitive inhibitor of Pim-1 kinase supplied as a high-purity research chemical. It inhibits Pim-1 (IC50 = 24 μM; Ki = 0.6 μM) and Pim-2 (IC50 = 100 μM), is provided as a white to off-white solid in a 10 mg package, and includes recommended storage conditions for powder and solutions.
  • Selective ATP-competitive Pim-1 kinase inhibitor suitable for research use.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a 10 mg solid for small-scale experiments.
  • Includes storage recommendations for powder and dissolved samples.
  • Contains chemical identifiers for unambiguous compound tracking.