Learn More
Medchemexpress LLC (Z)-10-Hydroxynortriptyline | 47132-19-4 | 99.7% | 279.38 | 1 MG
SDP
Catalog No. 5000365340
Encompass_Preferred
(Z)-10-Hydroxynortriptyline is a metabolite of Nortriptyline, a tricyclic antidepressant and the main active metabolite of Amitriptyline. It is used to relieve symptoms of depression and is intended for research use only.
  • Metabolite of Nortriptyline
  • Can be used to relieve symptoms of depression
  • Available as a white to off-white solid
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
  • Ships at room temperature within the continental US
Catalog No. 50-003-65340 Supplier Medchemexpress LLC Supplier No. 1MGHY100646A
(Z)-10-Hydroxynortriptyline is a metabolite of Nortriptyline, a tricyclic antidepressant and the main active metabolite of Amitriptyline. It is used to relieve symptoms of depression and is intended for research use only.
  • Metabolite of Nortriptyline
  • Can be used to relieve symptoms of depression
  • Available as a white to off-white solid
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
  • Ships at room temperature within the continental US