Learn More
Medchemexpress LLC (Z)-10-hydroxynortriptyline-d3 | 47132-19-4 | 99.0% | 282.40 | C19H18D3NO | 5 MG
SDP
Catalog No. 5000214181
Encompass_Preferred
Deuterium-labeled (Z)-10-hydroxynortriptyline-d3 is a stable isotope internal standard of 10-hydroxynortriptyline, provided as a high-purity analytical reagent for use as a tracer and quantitative reference in research applications. It is intended for analytical workflows requiring precise quantitation of metabolites and drug substances.
  • Stable isotope internal standard for accurate quantitation.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • High purity suitable for analytical assays and calibration.
  • Available in small research quantities for method development.
  • Useful as an internal standard or tracer in pharmacokinetic studies.
Catalog No. 50-002-14181 Supplier Medchemexpress LLC Supplier No. HY100646AS5MG
Deuterium-labeled (Z)-10-hydroxynortriptyline-d3 is a stable isotope internal standard of 10-hydroxynortriptyline, provided as a high-purity analytical reagent for use as a tracer and quantitative reference in research applications. It is intended for analytical workflows requiring precise quantitation of metabolites and drug substances.
  • Stable isotope internal standard for accurate quantitation.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
  • High purity suitable for analytical assays and calibration.
  • Available in small research quantities for method development.
  • Useful as an internal standard or tracer in pharmacokinetic studies.