Learn More
Medchemexpress LLC (Z)-11-octadecenal | 4273-95-4 | MFCD18642088 | 95.9% | 266.46 | C18H34O | 25mg
SDP
Catalog No. 5000270503
Encompass_Preferred
(Z)-11-Octadecenal is a female pheromone with diurnal stability isolated from the pheromone glands of female rice leaffolder moths[1]
Catalog No. 50-002-70503 Supplier Medchemexpress LLC Supplier No. HYW74282125MG
(Z)-11-Octadecenal is a female pheromone with diurnal stability isolated from the pheromone glands of female rice leaffolder moths[1]