Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-2-(4-((6-chloro-7-methyl-1H-indol-3-yl)methylene)-2,5-dioxoimidazolidin-1-yl)-2-(3,4-difluorophenyl) | 1416663-77-8 | >98.0% | 1 ML
SDP

Catalog No. 5000231844
Encompass_Preferred

RO-5963 is a small-molecule research inhibitor that disrupts p53 binding to MDM2 and MDMX, activating the p53 pathway. Reported IC50 values are approximately 17 nM (p53-MDM2) and 24 nM (p53-MDMX). The compound is supplied as a 10 mM solution in DMSO (1 mL) or as solids in multiple mg quantities for research use.

  • Dual inhibition of p53-MDM2 and p53-MDMX with low-nanomolar potency.
  • Available as a ready-to-use 10 mM solution in DMSO for cell-based assays.
  • Also available as solid packages for storage or custom formulation.
  • Molecular formula C24H21ClF2N4O5; molecular weight 518.90 g/mol.
  • Analytical purity typically ≥98% by HPLC where specified.

Catalog No. 50-002-31844 Supplier Medchemexpress LLC Supplier No. HY12008610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RO-5963 is a small-molecule research inhibitor that disrupts p53 binding to MDM2 and MDMX, activating the p53 pathway. Reported IC50 values are approximately 17 nM (p53-MDM2) and 24 nM (p53-MDMX). The compound is supplied as a 10 mM solution in DMSO (1 mL) or as solids in multiple mg quantities for research use.

  • Dual inhibition of p53-MDM2 and p53-MDMX with low-nanomolar potency.
  • Available as a ready-to-use 10 mM solution in DMSO for cell-based assays.
  • Also available as solid packages for storage or custom formulation.
  • Molecular formula C24H21ClF2N4O5; molecular weight 518.90 g/mol.
  • Analytical purity typically ≥98% by HPLC where specified.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.