Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Z 2-Cl -OSu, Advanced ChemTech

Catalog No. 01339355
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Catalog No. Quantity
01-339-355 1 g
01-339-354 25 g
01-339-356 100 g
3 options

Catalog No. 01-339-355

Supplier: Advanced Chem Tech RC80101G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65853-65-8
Molecular Formula C12H10ClNO5
Molecular Weight (g/mol) 283.66
InChI Key LVZHXYXNMHCBKC-UHFFFAOYSA-N
IUPAC Name (2-chlorophenyl)methyl 2,5-dioxopyrrolidin-1-yl carbonate
SMILES ClC1=CC=CC=C1COC(=O)ON1C(=O)CCC1=O

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Z 2-Cl -OSu
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.