Learn More
TARGETMOL CHEMICALS INC Z-2-decenoic acid 10MG
SDP
Catalog No. 502880335
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (Z)-2-decenoic acid (cis-dec-2-enoic acid) is an unsaturated short-chain fatty acid secreted by Pseudomonas aeruginosa. It can induce a dispersion response in biofilms formed by gram-negative and gram-positive bacteria and can also inhibit biofilm development.
Catalog No. 50-288-0335 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ025110MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (Z)-2-decenoic acid (cis-dec-2-enoic acid) is an unsaturated short-chain fatty acid secreted by Pseudomonas aeruginosa. It can induce a dispersion response in biofilms formed by gram-negative and gram-positive bacteria and can also inhibit biofilm development.