Learn More
Sigma Aldrich (Z)-3-CHLORO-3-(4-NITROPHENYL)ACRYLONITRILE
Catalog No. JRD05101G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
JRD05101G 1 g
1 options
Catalog No. JRD05101G Supplier Sigma Aldrich Supplier No. JRD05101G
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
  • Reactant for:
  • Stereoselective ketonization-olefination of indoles by co/mn-mediated oxidative cross-coupling of indoles via dioxygen activation
  • Enantioselective michael reaction
  • Segphos-ruthenium-catalyzed asymmetric hydrogenation
Specifications
Quantity 1 g