Learn More
Medchemexpress LLC Z-3578 | 473687-20-6 | 98.7% | 467.37 | C23H16Cl2N4OS | 10 MG
SDP
Catalog No. 5000295444
Encompass_Preferred
Z-3578 is a small-molecule antagonist of the Mas-related G protein-coupled receptor X2 (MrgX2) supplied for research use. It inhibits mast cell degranulation induced by substance P and compound 48/80 and displays anti-pseudoallergic activity with a reported KD of 729 nM.
  • Selective MrgX2 antagonist for in vitro pharmacology.
  • Demonstrated inhibition of mast cell degranulation induced by substance P and compound 48/80.
  • High purity suitable for research applications.
  • Well-characterized physical data including formula C23H16Cl2N4OS and molecular weight 467.37.
  • Stable under recommended storage when sealed and protected from moisture and light.
Catalog No. 50-002-95444 Supplier Medchemexpress LLC Supplier No. HY17245810MG
Z-3578 is a small-molecule antagonist of the Mas-related G protein-coupled receptor X2 (MrgX2) supplied for research use. It inhibits mast cell degranulation induced by substance P and compound 48/80 and displays anti-pseudoallergic activity with a reported KD of 729 nM.
  • Selective MrgX2 antagonist for in vitro pharmacology.
  • Demonstrated inhibition of mast cell degranulation induced by substance P and compound 48/80.
  • High purity suitable for research applications.
  • Well-characterized physical data including formula C23H16Cl2N4OS and molecular weight 467.37.
  • Stable under recommended storage when sealed and protected from moisture and light.