Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-4-Amino-4-oxobut-2-enoic acid | 557-24-4 | 96.9% | 1 ML
SDP

Catalog No. 5000342331
Encompass_Preferred

(Z)-4-Amino-4-oxobut-2-enoic acid is an endogenous metabolite. It is a white to off-white solid with a molecular weight of 115.09 and formula C4H5NO3. Classified as ketones, aldehydes, and acids, it is intended for research use only.

  • Endogenous metabolite
  • White to off-white solid
  • Molecular weight of 115.09
  • Formula C4H5NO3
  • Classified as ketones, aldehydes, and acids
  • For research use only

Catalog No. 50-003-42331 Supplier Medchemexpress LLC Supplier No. HY12875010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-4-Amino-4-oxobut-2-enoic acid is an endogenous metabolite. It is a white to off-white solid with a molecular weight of 115.09 and formula C4H5NO3. Classified as ketones, aldehydes, and acids, it is intended for research use only.

  • Endogenous metabolite
  • White to off-white solid
  • Molecular weight of 115.09
  • Formula C4H5NO3
  • Classified as ketones, aldehydes, and acids
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.