Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-4-Amino-4-oxobut-2-enoic acid | 557-24-4 | 96.9% | 5 G
SDP

Catalog No. 5000340583
Encompass_Preferred

(Z)-4-Amino-4-oxobut-2-enoic acid is an endogenous metabolite. This product is intended for research use only and is not for sale to patients.

  • Endogenous metabolite
  • Appears as a solid
  • Color is white to off-white
  • Molecular weight is 115.09
  • Molecular formula is C4H5NO3
  • Soluble in DMSO at 116.67 mg/mL
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-003-40583 Supplier Medchemexpress LLC Supplier No. HY1287505G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-4-Amino-4-oxobut-2-enoic acid is an endogenous metabolite. This product is intended for research use only and is not for sale to patients.

  • Endogenous metabolite
  • Appears as a solid
  • Color is white to off-white
  • Molecular weight is 115.09
  • Molecular formula is C4H5NO3
  • Soluble in DMSO at 116.67 mg/mL
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.