Learn More
Medchemexpress LLC (Z)-4-hydroxy-N'-((2-oxonaphthalen-1(2H)-ylidene)methyl)benzohydrazide | 2250156-71-7 | 99.0% | 306.32 g/mol | C18H14N2O3 | 5 MG
SDP
Catalog No. 5000222628
Encompass_Preferred
MEISi-2 is a small-molecule, selective inhibitor of MEIS transcription factors developed for research into hematopoietic stem cell self-renewal, cardiac injury, bone marrow transplantation, and cancer. Supplied as a high-purity research reagent, it is intended for in vitro screening and mechanistic studies.
  • Selective MEIS transcription factor inhibition for target validation.
  • High purity suitable for biochemical and cell-based assays.
  • Small pack size ideal for screening and pilot experiments.
  • Molecular weight 306.32 g/mol and formula C18H14N2O3 for accurate dosing calculations.
  • Supplied with CAS number 2250156-71-7 for unambiguous identification.
Catalog No. 50-002-22628 Supplier Medchemexpress LLC Supplier No. HY1342595MG
MEISi-2 is a small-molecule, selective inhibitor of MEIS transcription factors developed for research into hematopoietic stem cell self-renewal, cardiac injury, bone marrow transplantation, and cancer. Supplied as a high-purity research reagent, it is intended for in vitro screening and mechanistic studies.
  • Selective MEIS transcription factor inhibition for target validation.
  • High purity suitable for biochemical and cell-based assays.
  • Small pack size ideal for screening and pilot experiments.
  • Molecular weight 306.32 g/mol and formula C18H14N2O3 for accurate dosing calculations.
  • Supplied with CAS number 2250156-71-7 for unambiguous identification.