Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-4-hydroxy tamoxifen-d5 | 164365-20-2 | MFCD07369456 | 98.3% | C26H24D5NO2 | 10MG
SDP

Catalog No. 5000260859
Encompass_Preferred

(Z)-4-Hydroxy Tamoxifen-d5 is the deuterium-labeled (d5) analogue of (Z)-4-hydroxy tamoxifen provided as a high-purity analytical reagent for research use. It is intended for use as an internal standard, tracer, or analytical reference in quantitative mass spectrometry, metabolite profiling, and pharmacokinetic studies; handle and store according to recommended conditions to maintain stability.

  • Deuterium-labeled (d5) isotopologue for mass spectrometry applications.
  • High purity (98.3%) for reliable analytical performance.
  • Suitable as an internal standard or tracer in quantitative assays.
  • Chemical formula C26H24D5NO2; CAS 164365-20-2.
  • Available in small laboratory pack sizes for analytical workflows.
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C.

Catalog No. 50-002-60859 Supplier Medchemexpress LLC Supplier No. HY132477S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-4-Hydroxy Tamoxifen-d5 is the deuterium-labeled (d5) analogue of (Z)-4-hydroxy tamoxifen provided as a high-purity analytical reagent for research use. It is intended for use as an internal standard, tracer, or analytical reference in quantitative mass spectrometry, metabolite profiling, and pharmacokinetic studies; handle and store according to recommended conditions to maintain stability.

  • Deuterium-labeled (d5) isotopologue for mass spectrometry applications.
  • High purity (98.3%) for reliable analytical performance.
  • Suitable as an internal standard or tracer in quantitative assays.
  • Chemical formula C26H24D5NO2; CAS 164365-20-2.
  • Available in small laboratory pack sizes for analytical workflows.
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.