Learn More
Sigma Aldrich Fine Chemicals Biosciences (Z)-4-Hydroxytamoxifen, 68047-06-3, MFCD00278780, 5 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H79045MG
Encompass
Empirical Formula (Hill Notation): C26H29NO2, Molecular Weight: 387.51, Synonym: (Z)-4-OHT, (Z)-4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, trans-4-Hydroxytamoxifen, ≥98% Z isomer.
Catalog No. NC0707746