Learn More
BIOGEMS INTERNATIONAL INC (Z)-4-Hydroxytamoxifen 25mg
SDP
Catalog No. 501530233
Encompass
Tamoxifen is a modulator of the of the estrogen receptor, with tissue specific agonistic and antagonistic effects. (Z)-4-Hydroxytamoxifen is a Tamoxifen metabolite with increased potency and a 100x higher affinity for the estrogen receptors. It is also reported to inhibit lipid peroxidation and can be used to activate intein-linked or estrogen receptor ERT2 domain fused Cas9 to reduce off target effects in CRISPR-mediated gene editing.
Catalog No. 50-153-0233 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 680063725MG
Tamoxifen is a modulator of the of the estrogen receptor, with tissue specific agonistic and antagonistic effects. (Z)-4-Hydroxytamoxifen is a Tamoxifen metabolite with increased potency and a 100x higher affinity for the estrogen receptors. It is also reported to inhibit lipid peroxidation and can be used to activate intein-linked or estrogen receptor ERT2 domain fused Cas9 to reduce off target effects in CRISPR-mediated gene editing.