Learn More
Enzo Life Sciences (Z)-4-Hydroxytamoxifen (5mg). CAS: 68047-06-3, 65213-48-1
SDP
Supplier:  Enzo Life Sciences ALX550361M005
Encompass_Preferred
Metabolite of tamoxifen (ALX-550-095). Exhibits a more potent estrogen agonist/antagonist activity than the parent drug. Has antioxidant properties. Intramembranous inhibitor of lipid peroxidation. Alternative name: (Z)-4-[1-(p-(Dimethylaminoethoxy)phenyl)-2-phenyl-1-butenyl]phenol. Purity: ≥98% (UHPLC). Solubility: Soluble in 100% ethanol (20mg/ml) or methanol (10mg/ml); insoluble water. Long Term Storage: +4°C.
Catalog No. 50-200-9545