Learn More
STEMCELL Technologies (Z)-4-Hydroxytamoxifen, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 74052
Encompass
(Z)-4-Hydroxytamoxifen is a cell-permeable selective estrogen receptor modulator (SERM) that binds to estrogen receptors (ER) and estrogen-related receptors (ERR) with both estrogenic and anti-estrogenic effects (Poulin et al.). CANCER RESEARCH· Suppresses tumor growth in breast cancer cells (Freiss et al.; Osborne et al.).
Catalog No. 50-197-6170