Learn More
Medchemexpress LLC HY-19831A 5mg Medchemexpress, (Z)-4EGI-1 CAS:901787-88-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19831A5MG
Encompass_Preferred
Medchemexpress, HY-19831A 5mg (Z)-4EGI-1 CAS:901787-88-0 (Z)-4EGI-1 is the Z-isomer of 4EGI-1 and is an inhibitor of eIF4E/eIF4G interaction and of translation initiation. (Z)-4EGI-1 effectively binds to eIF4E with an IC50 of 43.5 μM and a Kd value of 8.74 μM. (Z)-4EGI-1 has anticancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0891