Learn More
Medchemexpress LLC (Z)-4EGI-1 | 901787-88-0 | 98.0% | 451.28 | 100 MG
SDP
Catalog No. 5000403396
Encompass_Preferred
(Z)-4EGI-1 is the Z-isomer of 4EGI-1, functioning as an inhibitor of eIF4E/eIF4G interaction and translation initiation. It effectively binds to eIF4E and has demonstrated anticancer activity by inhibiting cancer cell proliferation.
  • Inhibits eIF4E/eIF4G interaction
  • Inhibits translation initiation
  • Binds to eIF4E
  • Demonstrates anticancer activity
  • Reduces expressions of regulatory proteins
  • Inhibits cancer cell proliferation
Catalog No. 50-004-03396 Supplier Medchemexpress LLC Supplier No. HY19831A100MG
(Z)-4EGI-1 is the Z-isomer of 4EGI-1, functioning as an inhibitor of eIF4E/eIF4G interaction and translation initiation. It effectively binds to eIF4E and has demonstrated anticancer activity by inhibiting cancer cell proliferation.
  • Inhibits eIF4E/eIF4G interaction
  • Inhibits translation initiation
  • Binds to eIF4E
  • Demonstrates anticancer activity
  • Reduces expressions of regulatory proteins
  • Inhibits cancer cell proliferation