Learn More
Electron Microscopy Sciences Z-6040 Primer 100 ML
SDP
Catalog No. 50997415
Encompass_Preferred
This unique primer affords you a better adhesion between your resin and specimen that you are embedding and prevents the sample from pulling away from the resin during sectioning.
Catalog No. 50-997-415 Supplier Electron Microscopy Sciences Supplier No. 5044010
This unique primer affords you a better adhesion between your resin and specimen that you are embedding and prevents the sample from pulling away from the resin during sectioning.