Learn More
Medchemexpress LLC (Z)-9-propenyladenine | 1464851-21-5 | MFCD30718180 | 175.19 g/mol | C8H9N5 | 100 MG
SDP
Catalog No. 5000215691
Encompass_Preferred
(Z)-9-propenyladenine is the Z-isomer analytical standard of 9-propenyladenine used as a reference impurity for tenofovir disoproxil. It is supplied as a solid and as preprepared solutions in DMSO for analytical and method development use. Chemical formula C8H9N5; molecular weight 175.19 g/mol.
  • Used as an analytical reference standard for impurity identification and quantitation.
  • Supplied as solid and as 10 mM solutions in DMSO for convenience.
  • Suitable for HPLC and LC-MS method development and validation.
  • White to off-white solid appearance for visual inspection.
  • Chemical formula and molecular weight provided for accurate calculations.
Catalog No. 50-002-15691 Supplier Medchemexpress LLC Supplier No. HY100079A100MG
(Z)-9-propenyladenine is the Z-isomer analytical standard of 9-propenyladenine used as a reference impurity for tenofovir disoproxil. It is supplied as a solid and as preprepared solutions in DMSO for analytical and method development use. Chemical formula C8H9N5; molecular weight 175.19 g/mol.
  • Used as an analytical reference standard for impurity identification and quantitation.
  • Supplied as solid and as 10 mM solutions in DMSO for convenience.
  • Suitable for HPLC and LC-MS method development and validation.
  • White to off-white solid appearance for visual inspection.
  • Chemical formula and molecular weight provided for accurate calculations.