Learn More
Medchemexpress LLC (Z)-9-Tetradecenyl acetate | 16725-53-4 | 254.41 | 250 MG
SDP
Catalog No. 5000389579
Encompass_Preferred
Prodenialure A, also known as (Z)-9-Tetradecenyl acetate, is an ester product that appears as a colorless to light yellow liquid. It is initially sourced from animals, specifically the fall armyworm moth.
  • Ester product.
  • Colorless to light yellow liquid appearance.
  • Store pure form at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Sourced from fall armyworm moth.
Catalog No. 50-003-89579 Supplier Medchemexpress LLC Supplier No. 250MGHYN11582
Prodenialure A, also known as (Z)-9-Tetradecenyl acetate, is an ester product that appears as a colorless to light yellow liquid. It is initially sourced from animals, specifically the fall armyworm moth.
  • Ester product.
  • Colorless to light yellow liquid appearance.
  • Store pure form at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Sourced from fall armyworm moth.