Learn More
Enzo Life Sciences Z-AAD-CMK (1mg)
SDP
Catalog No. 502011270
Encompass_Preferred
Irreversible, cell-permeable inhibitor of granzyme B (ID50=0.3 µM). Does not inhibit granzyme A. Inhibition of granzyme B with Z-AAD-CMK prevented caspase-3 activation in T cells. Alternative name: Granzyme B inhibitor. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO (25mg/ml), 100% ethanol (25mg/ml) or water (20mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1270 Supplier Enzo Life Sciences Supplier No. BMLP1650001
Irreversible, cell-permeable inhibitor of granzyme B (ID50=0.3 µM). Does not inhibit granzyme A. Inhibition of granzyme B with Z-AAD-CMK prevented caspase-3 activation in T cells. Alternative name: Granzyme B inhibitor. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO (25mg/ml), 100% ethanol (25mg/ml) or water (20mg/ml). Long Term Storage: -20°C.