Learn More
Medchemexpress LLC (Z)-aconitic acid | 585-84-2 | MFCD00063184 | 99.3% | 174.11 g/mol | C6H6O6 | 5 MG
SDP
Catalog No. 5000301996
Encompass_Preferred
(Z)-aconitic acid is a small-molecule endogenous metabolite (cis-aconitic acid) reported to act as a glutamate decarboxylase inhibitor with activity that includes reduction of IκB-α phosphorylation and inhibition of inflammatory responses; supplied as a solid for research use.
  • Molecular formula C6H6O6 and molecular weight 174.11 g/mol.
  • High reported purity (99.28%).
  • Reported activity as a glutamate decarboxylase inhibitor and modulator of IκB-α phosphorylation.
  • Applicable to research on inflammatory and metabolic pathways.
  • Powder stability: -20°C for long-term storage; defined solvent storage conditions available.
Catalog No. 50-003-01996 Supplier Medchemexpress LLC Supplier No. HYW0168145MG
(Z)-aconitic acid is a small-molecule endogenous metabolite (cis-aconitic acid) reported to act as a glutamate decarboxylase inhibitor with activity that includes reduction of IκB-α phosphorylation and inhibition of inflammatory responses; supplied as a solid for research use.
  • Molecular formula C6H6O6 and molecular weight 174.11 g/mol.
  • High reported purity (99.28%).
  • Reported activity as a glutamate decarboxylase inhibitor and modulator of IκB-α phosphorylation.
  • Applicable to research on inflammatory and metabolic pathways.
  • Powder stability: -20°C for long-term storage; defined solvent storage conditions available.