Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-aconitic acid | 585-84-2 | MFCD00063184 | 99.3% | 174.11 g/mol | C6H6O6 | 50 MG
SDP

Catalog No. 5000301225
Encompass_Preferred

(Z)-Aconitic acid (cis-aconitic acid) is a research-grade organic acid (CAS 585-84-2) described as a glutamate decarboxylase inhibitor that reduces IκB-α phosphorylation. It is supplied in small quantities for biochemical and pharmacological research and is intended for laboratory research use only.

  • High purity: 99.28%.
  • Molecular weight: 174.11 g/mol.
  • Chemical formula: C6H6O6.
  • Available in small research packages including a 50 mg size.
  • Solid, white to light yellow appearance.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-003-01225 Supplier Medchemexpress LLC Supplier No. HYW01681450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-Aconitic acid (cis-aconitic acid) is a research-grade organic acid (CAS 585-84-2) described as a glutamate decarboxylase inhibitor that reduces IκB-α phosphorylation. It is supplied in small quantities for biochemical and pharmacological research and is intended for laboratory research use only.

  • High purity: 99.28%.
  • Molecular weight: 174.11 g/mol.
  • Chemical formula: C6H6O6.
  • Available in small research packages including a 50 mg size.
  • Solid, white to light yellow appearance.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.