Learn More
Medchemexpress LLC Z-aconitic acid (cis-aconitic acid) | 585-84-2 | MFCD00063184 | 99.3% | 174.11 g·mol⁻¹ | C6H6O6 | 10 MG
SDP
Catalog No. 5000300594
Encompass_Preferred
(Z)-Aconitic acid (CAS 585-84-2) is a research-grade analytical standard and biochemical probe. It is reported to act as a glutamate decarboxylase inhibitor and to reduce IκB-α phosphorylation. The material is supplied in solid and solution formats for laboratory research use only.
  • High purity suitable for analytical applications.
  • Available in multiple pack sizes, including small solid quantities and ready-to-use solutions.
  • Characterized molecular formula and molecular weight for accurate dosing.
  • Provided with purity documentation to support quality control.
  • Intended for laboratory research use only.
Catalog No. 50-003-00594 Supplier Medchemexpress LLC Supplier No. HYW016814R10MG
(Z)-Aconitic acid (CAS 585-84-2) is a research-grade analytical standard and biochemical probe. It is reported to act as a glutamate decarboxylase inhibitor and to reduce IκB-α phosphorylation. The material is supplied in solid and solution formats for laboratory research use only.
  • High purity suitable for analytical applications.
  • Available in multiple pack sizes, including small solid quantities and ready-to-use solutions.
  • Characterized molecular formula and molecular weight for accurate dosing.
  • Provided with purity documentation to support quality control.
  • Intended for laboratory research use only.