Learn More
Medchemexpress LLC HY-136626 1mg Medchemexpress, Z-Ala-Ala-Asn-AMC CAS:149697-16-5 Purity:>98%
SDP
Catalog No. 502030461
Encompass_Preferred
Medchemexpress, HY-136626 1mg Z-Ala-Ala-Asn-AMC CAS:149697-16-5 Z-Ala-Ala-Asn-AMC (Cbz-Ala-Ala-Asn-AMC) is the legumain substrate. Overexpressed legumain in 293 HEK-Leg cells potently cleaved CBZ-Ala-Ala-Asn-AMC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0461 Supplier Medchemexpress LLC Supplier No. HY1366261MG
Medchemexpress, HY-136626 1mg Z-Ala-Ala-Asn-AMC CAS:149697-16-5 Z-Ala-Ala-Asn-AMC (Cbz-Ala-Ala-Asn-AMC) is the legumain substrate. Overexpressed legumain in 293 HEK-Leg cells potently cleaved CBZ-Ala-Ala-Asn-AMC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.