Learn More
Bachem Z-Ala-Ala-Asn-AMC
SDP
Catalog No. 50260351
Encompass_Preferred
Z-Ala-Ala-Asn-AMC, 149697-16-5, C28H31N5O8, Mr 565.58, 50mg. Z-AAN-AMC, standard substrate for legumain (EC 3.4.22.34), was originally used for the plant enzyme and later to discover the mammalian form of the enzyme. Legumain may be a physiologic local regulator of human osteoclast (OCL) activity that can negatively modulate OCL.
Catalog No. 50-260-351 Supplier Bachem Supplier No. 4033201.0050
Z-Ala-Ala-Asn-AMC, 149697-16-5, C28H31N5O8, Mr 565.58, 50mg. Z-AAN-AMC, standard substrate for legumain (EC 3.4.22.34), was originally used for the plant enzyme and later to discover the mammalian form of the enzyme. Legumain may be a physiologic local regulator of human osteoclast (OCL) activity that can negatively modulate OCL.