Learn More
Carbosynth LLC. Z-Ala-Ala-Leu-pNA, 0.1g, MW: 527.57, Molecular formula: C26H33N5O7, CAS# 61043-33-2
SDP
Supplier:  Carbosynth LLC. SAP31270.1G
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Alanyl-L-Alanyl-L-Leucine p-Nitroanilide, Application: Substrate for Substilisin A and Serine Protease of Bacillus substilis IFO3027/Neprilysin, Storage Temperature: 2-10C, References: V.M. Stepanov, A.Y. Strongin, L.S. Izotova, Z.T. Abramov, L.A. Lyublinskaya, L.M. Ermakova, L.A. Baratova, and L.P. Belyanova, Biochem. Biophys. Res. Commun., 77, 298 (1977), Y. Shimizu, T. Nishino, and S. Murao, Agric. Biol. Chem., 47, 1775 (1983), Y. Takaki, N. Iwata, S. Tsubuki, S. Taniguchi, S. Toyoshima, B. Lu, N.P. Gerard, C. Gerard, H.-J. Lee, K. Shirotani, and T.C. Saido, J. Biochem., 128, 897 (2000)
Catalog No. 50-168-7026