Learn More
Medchemexpress LLC Z-Ala-Arg-Arg-AMC | 90468-18-1 | 98.4% | 692.77 g/mol | C33H44N10O7 | 5 MG
SDP
Catalog No. 5000221874
Encompass_Preferred
Z-Ala-Arg-Arg-AMC is a fluorogenic peptide substrate for quantifying trypsin-like activity of the proteasome. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescence signal used in biochemical and enzymatic assays.
  • Generates a fluorescent AMC signal upon proteolytic cleavage.
  • Designed for measuring trypsin-like proteasome activity.
  • High purity (~98.4%) suitable for research assays.
  • Supplied as a solid; soluble in DMSO at about 50 mg/mL with ultrasonic assistance.
  • Store sealed, away from moisture, under recommended low-temperature conditions.
  • Available in small research-scale quantities (mg scale).
Catalog No. 50-002-21874 Supplier Medchemexpress LLC Supplier No. HY1349615MG
Z-Ala-Arg-Arg-AMC is a fluorogenic peptide substrate for quantifying trypsin-like activity of the proteasome. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescence signal used in biochemical and enzymatic assays.
  • Generates a fluorescent AMC signal upon proteolytic cleavage.
  • Designed for measuring trypsin-like proteasome activity.
  • High purity (~98.4%) suitable for research assays.
  • Supplied as a solid; soluble in DMSO at about 50 mg/mL with ultrasonic assistance.
  • Store sealed, away from moisture, under recommended low-temperature conditions.
  • Available in small research-scale quantities (mg scale).