Learn More
Medchemexpress LLC Z-ala-glu-val-asp fluoromethyl ketone | 1135688-47-9 | >95.0% | 610.63 | C28H39FN4O10 | 25MG
SDP
Catalog No. 5000237957
Encompass_Preferred
Z-AEVD-FMK is a cell-permeable peptide fluoromethyl ketone that irreversibly inhibits caspase-10, preventing Bid activation and the mitochondrial release of apoptosis-inducing factor (AIF) to reduce apoptosis in cellular models. The compound is supplied as a crystalline solid, has molecular formula C28H39FN4O10, molecular weight 610.63, and is typically provided at ≥95% purity. Handle protected from light and store under nitrogen; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
  • Irreversible caspase-10 inhibition for apoptosis pathway studies.
  • Cell-permeable peptide fluoromethyl ketone scaffold.
  • High purity suitable for biochemical assays and cell experiments.
  • Stable crystalline form for convenient storage and handling.
  • Storage instructions provided to preserve compound integrity.
Catalog No. 50-002-37957 Supplier Medchemexpress LLC Supplier No. HY12530525MG
Z-AEVD-FMK is a cell-permeable peptide fluoromethyl ketone that irreversibly inhibits caspase-10, preventing Bid activation and the mitochondrial release of apoptosis-inducing factor (AIF) to reduce apoptosis in cellular models. The compound is supplied as a crystalline solid, has molecular formula C28H39FN4O10, molecular weight 610.63, and is typically provided at ≥95% purity. Handle protected from light and store under nitrogen; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
  • Irreversible caspase-10 inhibition for apoptosis pathway studies.
  • Cell-permeable peptide fluoromethyl ketone scaffold.
  • High purity suitable for biochemical assays and cell experiments.
  • Stable crystalline form for convenient storage and handling.
  • Storage instructions provided to preserve compound integrity.