Learn More
Bachem Z-Arg-Arg-AMC
SDP
Catalog No. 50260283
Encompass_Preferred
Z-Arg-Arg-AMC Hydrochloride, 88937-61-5, C30H39N9O6, Mr 621.70, 50mg. Z-RR-AMC, an excellent, specific substrate for cathepsin B, an enzyme found in secretory vesicles. Cathepsin B has been identified as a b-secretase, as such it is involved in b-amyloid production. Z-RR-AMC is a convenient fluorogenic substrate for stem bromelain, as it is scarcely affected by fruit bromelain.
Catalog No. 50-260-283 Supplier Bachem Supplier No. 4004789.0050
Z-Arg-Arg-AMC Hydrochloride, 88937-61-5, C30H39N9O6, Mr 621.70, 50mg. Z-RR-AMC, an excellent, specific substrate for cathepsin B, an enzyme found in secretory vesicles. Cathepsin B has been identified as a b-secretase, as such it is involved in b-amyloid production. Z-RR-AMC is a convenient fluorogenic substrate for stem bromelain, as it is scarcely affected by fruit bromelain.