Learn More
Medchemexpress LLC Z-Arg-Arg-AMC hydrochloride | 136132-67-7 | MFCD00133576 | 99.8% | 658.15 | C30H40ClN9O6 | 5 MG
SDP
Catalog No. 5000223461
Encompass_Preferred
Z-Arg-Arg-AMC hydrochloride is a fluorogenic peptide substrate selective for cathepsin B. Cleavage releases 7-amido-4-methylcoumarin (AMC), producing a fluorescent signal used in sensitive in vitro protease activity assays and kinetic studies. Intended for research use only.
  • Selective substrate for cathepsin B.
  • Fluorogenic AMC reporter enables sensitive fluorescence-based detection.
  • Suitable for kinetic and endpoint protease assays.
  • Available in small sizes for assay development and screening.
  • High purity (≈99.8%) for reproducible results.
Catalog No. 50-002-23461 Supplier Medchemexpress LLC Supplier No. HY1344345MG
Z-Arg-Arg-AMC hydrochloride is a fluorogenic peptide substrate selective for cathepsin B. Cleavage releases 7-amido-4-methylcoumarin (AMC), producing a fluorescent signal used in sensitive in vitro protease activity assays and kinetic studies. Intended for research use only.
  • Selective substrate for cathepsin B.
  • Fluorogenic AMC reporter enables sensitive fluorescence-based detection.
  • Suitable for kinetic and endpoint protease assays.
  • Available in small sizes for assay development and screening.
  • High purity (≈99.8%) for reproducible results.