Learn More
Carbosynth LLC. Z-Arg-Arg-MCA (5 mg vial), 1ea, MW: 621.69, Molecular formula: C30H39N9O6, CAS# 88937-61-5
SDP
Supplier:  Carbosynth LLC. MRR3123V5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Arginyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Z-Arg-Arg-AMC, Z-RR-AMC, Z-RR-MCA, Purity: , Grade A, Application: Substrate for Cathepsin B, Salt Form:, Hydrochloride Form, Storage Temperature: -20C, References: A.J. Barrett and H. Kirschke, In, Proteolytic Enzymes Part C, Methods in Enzymology, Vol. 80, (L. Lorand, Ed.), Academic Press, New York, 1981, pp. 535.
Catalog No. 50-168-6020