Learn More
Medchemexpress LLC Z-Arg-Arg-pNA | 122630-71-1 | 99.17% | 584.63 | 1 MG
SDP
Catalog No. 5000396677
Encompass_Preferred
Z-Arg-Arg-pNA is a substrate for cathepsin B, designed to detect the activity of this enzyme. This product is provided as a solid, appearing white to off-white, and is soluble in DMSO.
  • Substrate for cathepsin B
  • Can be used to detect enzyme activity
  • Solid appearance, white to off-white
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-003-96677 Supplier Medchemexpress LLC Supplier No. HYP45001MG
Z-Arg-Arg-pNA is a substrate for cathepsin B, designed to detect the activity of this enzyme. This product is provided as a solid, appearing white to off-white, and is soluble in DMSO.
  • Substrate for cathepsin B
  • Can be used to detect enzyme activity
  • Solid appearance, white to off-white
  • Soluble in DMSO (100 mg/mL)