Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Arg-Arg-pNA | 122630-71-1 | 99.2% | 584.63 | 10 MG
SDP

Catalog No. 5000398802
Encompass_Preferred

Z-Arg-Arg-pNA is a substrate for cathepsin B and can be used to detect this enzyme activity. It is for research use only and not sold to patients.

  • Functions as a substrate for cathepsin B
  • Utilized for detecting enzyme activity
  • Purity of 99.2%
  • Appears as a white to off-white solid
  • Molecular weight of 584.63
  • Formula: C26H36N10O6
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-98802 Supplier Medchemexpress LLC Supplier No. HYP450010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Arg-Arg-pNA is a substrate for cathepsin B and can be used to detect this enzyme activity. It is for research use only and not sold to patients.

  • Functions as a substrate for cathepsin B
  • Utilized for detecting enzyme activity
  • Purity of 99.2%
  • Appears as a white to off-white solid
  • Molecular weight of 584.63
  • Formula: C26H36N10O6
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.