Learn More
Bachem Z-Arg-Arg-pNA · 2 HCl
SDP
Catalog No. 50260390
Encompass_Preferred
Z-Arg-Arg-pNA · 2 HCl, 201807-90-1, C26H36N10O6 · 2 HCl, Mr 657.56, 50mg. Z-RR-pNA, chromogenic substrate for cathepsin B and stem bromelain.
Catalog No. 50-260-390 Supplier Bachem Supplier No. 4008026.0050
Z-Arg-Arg-pNA · 2 HCl, 201807-90-1, C26H36N10O6 · 2 HCl, Mr 657.56, 50mg. Z-RR-pNA, chromogenic substrate for cathepsin B and stem bromelain.